Product Name
Kolliphor® P 407 oxyethylene 71.5-74.9 %
Product Number
62035
Product Brand
sigma
CAS Number
9003-11-6
Molecular Weight
Test
Specification
APPEARANCE (COLOR)REPORT RESULT
APPEARANCE (FORM)REPORT RESULT
COLOR (IN APHA)≤ 120 APHA (50/50 IN CH3OH)
REMARKS ON GCPROPYLENE OXIDE ≤ 5 PPM
SOLUBILITY (METHOD)10% AQUEOUS SOLUTION
SOLUBILITYCOLORLESS, CLEAR
PH-TEST5.0 - 7.5 (1:40)
WATER≤ 1.0 %
RESIDUE ON IGNITION≤ 0.30 %
REM. ON PHYSICAL DATAAVERAGE MOLECULAR WEIGHT 9840 - 14600 G/MOL
INFRARED SPECTRUMCONFORMS TO STRUCTURE
PHARMACOPOEA TESTSBUTYLHYDROXYTOLUENE 50 - 125 PPM
OXYETHYLENE 71.5 - 74.9 %
APPLICATION TESTCLOUD POINT TEMPERATURE (10 G/L IN H2O) ≥ 100 DEG C
CONGEALING POINT 50 - 62 DEG C
IDENTIFICATION AIDENTIFICATION MUST CONFORM
DIOXAN≤ 5 ppm
ETHYLENE GLYCOL AND DIETHYLENE GLYCOLSUM ≤ 0.25 %
ETHYLENE OXIDE≤ 1 ppm
ARSENIC≤ 2 ppm
HEAVY METALS≤ 20 ppm
TOTAL ASH≤ 0.4 %
PROOF OF IDENTITYUNSATURATION 0.031 - 0.065 MEQ/G
MISCELLANEOUS TESTSPURITY (ACID) MUST COMPLY