MilliporeSigma
GC Analysis of Aldehydes (PFBOA Derivatives) in Beer on SLB®-5ms after SPME using a 65 μm PDMS/DVB Fiber
Materials
Related Product
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Used Together
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
CONDITIONS
sample preparationderivatization procedure: Add 100 μL of PFBOA solution (6 g/L) to 10 mL of deionized water in a 20 mL glass vial and seal. (Insert the PDMS/DVB SPME fiber in the headspace of the PFBOA solution for 10 min at 50 °C.)
sample/matrix10 mL of degassed beer, 3.5 gm sodium chloride in 20 mL vial
SPME fiberPDMS-DVB, 23 guage, 65 μm, Auto, Pk/3 (57345-U)
extraction60 min, headspace, 50 °C, agitation (5 min on, 30 min off)
desorption process0.10 min, 250 °C
columnSLB-5ms, 30 m x 0.25 mm I.D., 0.50 μm (28473-U)
oven40 °C, 7 °C/min to 250 °C (14 min)
inj. temp.250 °C
detectorMSD, SIM
MSD interface275 °C
carrier gashelium, 1.1 mL/min
injectionsplitless
liner0.75 mm I.D. SPME liner
sampleBeer
Description
Analysis NoteThe formation of aldehydes is a major contributor to the deterioration in flavor or "staling" of beer upon storage. Research indicates that the source of these aldehydes is the type of malt used and quality of the wort produced during the brewing process. This application illustrates the use of SPME to extract aldehydes from beer, followed by on-fiber derivatization with 0-(2,3,4,5,6- pentafluorobenzyl)hydroxylamine hydrochloride (PFBOA). The aldehydes are then analyzed by GC/MS-SIM as their corresponding PFBOA derivatives. The aldehydes exist as geometric isomers, resulting in two derivatives for each. In the case of several of the analytes, these two derivatives were resolved chromatographically.
Legal InformationSLB is a registered trademark of Merck KGaA, Darmstadt, Germany