manufacturer/tradename
WPI
General description
FLOX is a device for measuring fluorescence lifetime, phase and intensity. It uses LED excitation and photodiode detection with filter-based wavelength selection for easy experimental set-up and control.
FLOX is especially useful for oxygen sensing applications where stability and sensitivity to drift is important and where sample set-ups must be left undisturbed for long periods of time.
Elija entre una de las versiones más recientes:
¿Ya tiene este producto?