Skip to Content
Merck

BDPH9BCDA5BC

Bide Pharmatech

2-(Allylamino)nicotinic acid

Synonym(s):

2-(allylamino)nicotinic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H10O2N2
CAS Number:
Molecular Weight:
178.19
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H10N2O2/c1-2-5-10-8-7(9(12)13)4-3-6-11-8/h2-4,6H,1,5H2,(H,10,11)(H,12,13)

Inchi Key

WDIHOHTUZOCWRQ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 2-(Allylamino)nicotinic acid




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library