Skip to Content
Merck

SML2937

iHAP1

≥98% (HPLC)

Synonym(s):

(2-Chloro-10H-phenothiazin-10-yl)(4-methoxyphenyl)methanone, 2-Chloro-10-(4-methoxybenzoyl)-10H-phenothiazine

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

Empirical Formula (Hill Notation):
C20H14ClNO2S
CAS Number:
Molecular Weight:
367.85
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

S1c2c(cc(cc2)Cl)N(c4c1cccc4)C(=O)c3ccc(cc3)OC

InChI key

CSWHYHPUEDNIQY-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
T8516E5406SML2065
form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥99%

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

-

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

solubility

DMSO: 2 mg/mL, clear

solubility

ethanol: soluble 5 mg/mL, H2O: soluble 50 mg/mL, clear, colorless to yellow, DMSO: soluble

solubility

DMSO: >5 mg/mL at ~60 °C, clear

solubility

DMSO: 2 mg/mL, clear

color

white to beige

color

white to off-white

color

white

color

white to beige

Biochem/physiol Actions

iHAP1 is a potent activator of protein phosphatase 2A (PP2A) that allosterically facilitates assembly and activation of PP2A holoenzyme complex consisting PPP2R1A, PPP2R5E (B56ε, regulatory), and PPP2CA subunits. iHAP1 induces dephosphorylation of MYBL2 transcription factor on Ser241, causing irreversible cell cycle arrest of leukemia and other cancer cells in prometaphase. iHAP1 exhibits significant antitumor activity and lack of toxicity in T-ALL xenograft mice models. Normal CD34+ HSPCs are appears to be resistant to iHAP1. iHAP1 does not inhibit dopamine receptor D2.
potent allosteric activator of protein phosphatase 2A (PP2A) containing regulatory subunit PPP2R5E (B56ε)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ken Morita et al.
Cell, 181(3), 702-715 (2020-04-22)
Protein phosphatase 2A (PP2A) enzymes can suppress tumors, but they are often inactivated in human cancers overexpressing inhibitory proteins. Here, we identify a class of small-molecule iHAPs (improved heterocyclic activators of PP2A) that kill leukemia cells by allosterically assembling a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service