Skip to Content
Merck

ENA311759159

Enamine

2-chloro-1-[2-(4-fluorophenyl)morpholin-4-yl]ethan-1-one

Synonym(s):

4-(chloroacetyl)-2-(4-fluorophenyl)morpholine

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

CAS Number:
Molecular Weight:
257.69
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C12H13ClFNO2/c13-7-12(16)15-5-6-17-11(8-15)9-1-3-10(14)4-2-9/h1-4,11H,5-8H2

Inchi Key

WYWAGVSOZFMDHC-UHFFFAOYSA-N

Storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service