Merck
  • Several types of adherent cells elaborate a dialyzable lymphopoietic cofactor (Abelson Growth Promoter).
Several types of adherent cells elaborate a dialyzable lymphopoietic cofactor (Abelson Growth Promoter).
International journal of cell cloning (1990-11-01)
M Muirhead, R Davis, R C Schwartz, T J Waldschmidt, L Ackermann, G Palumbo, R G Smith
ABSTRACT
We have cloned a stromal cell from mouse bone marrow selected on the basis of its ability to promote the growth of an Abelson virus-transformed pre-B cell line. These stromal cells have smooth muscle features, and the ultrafiltrate of stromal cell-conditioned medium has proliferative effects on all feeder layer-responsive mouse pre-B cell lines tested, as well as on normal pre-B cells. One of these low MW substances is a protease-resistant factor with an MW of approximately 450 Da which we designate Abelson Growth Promoter (AGP). AGP was initially characterized as an activity which promotes the growth of Abelson virus-transformed mouse pre-B cells, but it also promotes the growth of a ras-transformed pre-B cell line as a single agent and in synergistic fashion with recombinant interleukin (IL) 7. AGP as a single agent has no effect on normal pre-B cells, which have a brisk response to IL-7. In contrast, transformed pre-B cells display a blunted response to IL-7. We propose that AGP plays a role in normal lymphopoiesis by expanding clones of pre-B cells which have been activated by other stromal cell-derived signals, such as IL-7, and directly promotes the growth of nascently transformed pre-B cells.
Sigma-Aldrich
Sephadex® G-10, Medium
Pricing and availability is not currently available.