Merck
  • A hydrophilic immobilized trypsin reactor with N-vinyl-2-pyrrolidinone modified polymer microparticles as matrix for highly efficient protein digestion with low peptide residue.
A hydrophilic immobilized trypsin reactor with N-vinyl-2-pyrrolidinone modified polymer microparticles as matrix for highly efficient protein digestion with low peptide residue.
Journal of chromatography. A (2012-03-27)
Hao Jiang, Huiming Yuan, Yu Liang, Simin Xia, Qun Zhao, Qi Wu, Lihua Zhang, Zhen Liang, Yukui Zhang
ABSTRACT
In this work, a novel kind of N-vinyl-2-pyrrolidinone (NVP) modified poly acrylic ester microspheres was prepared, followed by trypsin immobilization to prepare a hydrophilic immobilized enzyme reactor (IMER), to achieve highly efficient protein digestion with low peptide residue. The nonspecific adsorption of peptides on such an IMER was evaluated by the in sequence digestion of bovine serum albumin (BSA) and myoglobin. Without NVP modification, both proteins could be identified after digestion by a 5 cm-length IMER, but 18 peptides of BSA were found in the digests of myoglobin caused by the nonspecific adsorption of the matrix. With NVP modification, the hydrophilicity of IMER was greatly improved, resulting in not only the sequence coverage of myoglobin increased from 63% to 73%, but also no residual peptides from BSA observed in myoglobin digests. Although the sequence coverages of proteins obtained by the IMER were comparable to those obtained by in-solution digestion, the digestion time was shortened from 24h to 1 min. By such an IMER, a protein mixture, containing BSA, myoglobin, and cytochrome c (100, 1 and 0.01 μg/mL, respectively), was digested, and all proteins were unambiguously identified with improved sequence coverages than that achieved by in-solution digestion. Furthermore, the hydrophilic IMER was also off-line coupled to nano-RPLC-ESI-MS/MS for the analysis of proteins extracted from yeast. After 1.5 min digestion, 271 protein groups with at least 2 distinct peptides were identified, much more than those obtained by 24h in-solution digestion (192 protein groups), indicating the great potential of such an IMER for proteome analysis.
Sigma-Aldrich
1-Vinyl-2-pyrrolidinone, contains sodium hydroxide as inhibitor, ≥99%