- Novel microbial metabolites of the phoslactomycins family induce production of colony-stimulating factors by bone marrow stromal cells. I. Taxonomy, fermentation and biological properties.
Novel microbial metabolites of the phoslactomycins family induce production of colony-stimulating factors by bone marrow stromal cells. I. Taxonomy, fermentation and biological properties.
The Journal of antibiotics (1993-10-01)
T Kohama, R Enokita, T Okazaki, H Miyaoka, A Torikata, M Inukai, I Kaneko, T Kagasaki, Y Sakaida, A Satoh
PMID7503975
ABSTRACT
Three metabolites were isolated from the culture broth of an actinomycete strain identified as Streptomyces platensis SANK 60191, that induce the production of colony-stimulating factors (CSFs) by stromal cell line KM-102 at ED50 concentrations from 40 to 200 ng/ml. The compounds induced quantities of granulocyte CSF (G-CSF) and granulocyte-macrophage CSF (GM-CSF) comparable to those induced by interleukin-1, a strong CSF inducer. These metabolites were called leustroducsins (A, B and C) and were later found to be structurally related to phoslactomycins. This is the first report of CSF inducing activity by members of the phoslactomycin class.