Merck
  • Backbone ¹H, ¹³C, ¹⁵N NMR assignments of yeast OMP synthase in unliganded form and in complex with orotidine 5'-monophosphate.
Backbone ¹H, ¹³C, ¹⁵N NMR assignments of yeast OMP synthase in unliganded form and in complex with orotidine 5'-monophosphate.
Biomolecular NMR assignments (2013-01-15)
Michael Riis Hansen, Richard Harris, Eric W Barr, Hong Cheng, Mark E Girvin, Charles Grubmeyer
ABSTRACT
The type I phosphoribosyltransferase OMP synthase (EC 2.4.2.10) is involved in de novo synthesis of pyrimidine nucleotides forming the UMP precursor orotidine 5'-monophosphate (OMP). The homodimeric enzyme has a Rossman α/β core topped by a base-enclosing "hood" domain and a flexible domain-swapped catalytic loop. High-resolution X-ray structures of the homologous Salmonella typhimurium and yeast enzymes show that a general compacting of the core as well as movement of the hood and a major disorder-to-order transition of the loop occur upon binding of ligands MgPRPP and orotate. Here we present backbone NMR assignments for the unliganded yeast enzyme (49 kDa) and its complex with product OMP. We were able to assign 212-213 of the 225 non-proline backbone (15)N and amide proton resonances. Significant difference in chemical shifts of the amide cross peaks occur in regions of the structure that undergo movement upon ligand occupancy in the S. typhimurium enzyme.
Product Description
Millipore
HIS-Select® Cobalt Affinity Gel, (1:1 suspension in a 30% ethanol solution)
Pricing and availability is not currently available.