Millipore Express® filters. Regional supply. Global quality.
We are expanding our presence in Europe, the Middle East, and Africa with additional manufacturing capacity. By establishing in-region production for Millipore Express® filters in Blarney, Ireland, we are strengthening our supply chain resilience with shorter shipping distances. All supported by best-in-class global quality standards.
Experience the benefits of Millipore Express® filters—click here to request your free sample.
Why Choose Millipore Express® Filters?
Millipore Express® filters are time-tested polyethersulfone (PES) membrane filters that can be used for any sterilizing filtration operation in biomanufacturing. This family of sterilizing-grade membrane filters delivers high flux, excellent filter capacity, and is offered with different pore sizes and formats to meet the needs of different unit operations. Be confident in your sterilizing filtration with Millipore Express® filters!
Key Benefits
- High capacity for improved process economics
- High flux enables faster processing
- Sterilizing-grade membranes for safety assurance
- Available in cartridge, irradiation-compatible capsules, and high-area formats for maximum flexibility
Millipore Express® filter family includes:
- Millipore Express® SHRP filters: for additional protection in upstream cell culture processes; high capacity, sterilizing-grade, mycoplasma removal 0.5/0.1 µm filters
- Millipore Express® SHC filters: for efficient filtration of plugging streams; high capacity, sterilizing-grade 0.5/0.2 µm filters
- Millipore Express® SHF filters: for critical sterile filtration applications; high flux, high throughput, sterilizing-grade 0.2 µm filters
- Millipore Express® PHF filters: for less critical sterile filtration applications; cost-effective high flux, high throughput, sterilizing-grade 0.2 µm filters
Check our Millipore Express® Family Guide to learn more
Backed by the Emprove® Program
Millipore Express® filters are supported by the Emprove® Program, which offers:
- Comprehensive documentation including product-specific quality data
- Support for qualification, risk assessment, and process optimization
- Data in easily accessible, ready-to-use dossiers
End-to-End Expertise
Millipore Express® filters are backed by decades of sterile processing experience and our technical teams can help optimize and then validate your process, reducing your risk and accelerating your path to market.
Let us help you streamline your workflow and boost confidence in your process.
Try Millipore Express® filters today.
Related Products
Response not successful: Received status code 500
Related Resources
- Millipore Express® Family Guide
Our Millipore Express® filters can be used in a wide range of applications including filtration of cell culture media and feeds and for sterile filtration at different steps in monoclonal antibodies (mAbs), vaccine, plasma, viral vector and large and small volume parenteral production.
- Application Note: Cell Culture Media Filtration. Selection and Sizing Filters for CHO Cell Culture Processes
Explore throughput performance of Millipore Express® sterilizing filters for cell culture media filtration and their impact on filter sizing.
- Application Note: Cell Culture Media Filtration. Evaluating Performance of CHO Cell Cultures
Learn how to check if your media filter impacts performance. Studies confirmed PES and PVDF membrane filters did not affect cell growth and viability.
- Prefilters and Sterilizing-grade Filter Formats: Selection Guide
Sterilizing filters are offered in disposable capsules and cartridge formats. This guide will help you find the right format for your bioburden control.
Get to Know the Products from Blarney
Experience the benefits of our filters for yourself. Request a free sample now by filling out the form below. Samples will be shipped from another factory until production in Blarney starts.
Blarney Products Trial Offer Official Rules. VALID ONLY in Austria, Belgium, Bulgaria, Croatia, Republic of Cyprus, Czech Republic, Denmark, Estonia, Finland, France, Germany, Greece, Hungary, Ireland, Italy, Latvia, Lithuania, Luxembourg, Malta, Netherlands, Norway, Poland, Portugal, Romania, Slovakia, Slovenia, Spain, Sweden, Switzerland and the UK.