EMAIL THIS PAGE TO A FRIEND
The international journal of biochemistry & cell biology
S-Adenosylmethionine.
PMID 10762064
Abstract
S-Adenosyl-Lmethionine (SAM) is an important molecule in normal cell function and survival. SAM is utilized by three key metabolic pathways: transmethylation; transsulfuration; and polyamine synthesis. In transmethylation reactions, the methyl group of SAM is donated to a large variety of acceptor substrates including DNA, phospholipids and proteins. Thus, interference of these reactions can affect a wide spectrum of processes ranging from gene expression to membrane fluidity. In transsulfuration, the sulfur atom of the SAM is converted via a series of enzymatic steps to cysteine, a precursor of taurine and glutathione, a major cellular anti-oxidant. Polyamines are required for normal cell growth. Given the importance of SAM in tissue function, it is not surprising that this molecule is being investigated as a possible therapeutic agent for the treatment of various clinical disorders.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9384
S-(5′-Adenosyl)-L-homocysteine, crystalline
C14H20N6O5S