EMAIL THIS PAGE TO A FRIEND
Archives of pharmacal research
Cytotoxic triterpenes from Crataegus pinnatifida.
PMID 10836742
Abstract
Bioassay-guided fractionation of Crataegus pinnatifida (Rosaceae) gave two cytotoxic ursane-type triterpenes which were identified as uvaol (1) and ursolic acid (2) by physicochemical and spectroscopic methods. 3-Oxo-ursolic acid (3) was synthesized from ursolic acid (2) by Jones method. The cytotoxic activities of these compounds were tested against murine L1210 and human cancer cell lines (A549, SK-OV-3, SK-MEL-2, XF498, and HCT15) in vitro. Compounds 1 and 2 showed moderate cytotoxicities against L1210, whereas they showed weak activities against human cancer cell lines. However, compound 3 exhibited potent cytotoxic activities both in murine and in human cancer cell lines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
U6628
Uvaol, ≥95%
C30H50O2