EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Synthesis and pharmacological evaluation of novel arylpiperazine derivatives as nonsteroidal androgen receptor antagonists.
PMID 15516756
Abstract
The search for novel antiandrogens by high-throughput screening (HTS) of the Yamanouchi chemical library led to the discovery of the lead compound (5), which possesses an arylmorpholine moiety. Through the optimization of the lead compound (5), we have found a series of novel arylpiperazine derivatives. Among them, 4-[4-cyano-(3-trifluoromethyl)phenyl]-N-(4-fluorophenyl)piperazine-1-carboxamide (22; YM-92088) exhibited a potent AR antagonistic activity with an IC(50) value of 0.47 microM in the reporter assay, which is more potent than bicalutamide (4) which has an IC(50) value of 0.89 microM.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
135151
3-Chloro-4′-fluoropropiophenone, 98%
C9H8ClFO