EMAIL THIS PAGE TO A FRIEND
Journal of the American Academy of Dermatology
Fixed drug eruption caused by dimenhydrinate.
PMID 21315964
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D2396
Dimenhydrinate
C17H21NO · C7H7ClN4O2