EMAIL THIS PAGE TO A FRIEND
Journal of pharmacological sciences
Novel action of the chalcone isoliquiritigenin as a cystic fibrosis transmembrane conductance regulator (CFTR) inhibitor: potential therapy for cholera and polycystic kidney disease.
PMID 22186622
Abstract
Overstimulation of cAMP-activated Cl(-) secretion can cause secretory diarrhea. Isoliquiritigenin (ISLQ) is a plant-derived chalcone that has a wide range of biological activities. The present study thus aimed to investigate the effect of ISLQ on cAMP-activated Cl(-) secretion in human intestinal epithelium, especially the underlying mechanism and therapeutic application. Short-circuit current analysis of human intestinal epithelial (T84) cell monolayers revealed that ISLQ dose-dependently inhibited cAMP-activated Cl(-) secretion with an IC(50) of approximately 20 µM. ISLQ had no effect on either basal short-circuit current or Ca(2+)-activated Cl(-) secretion. Apical Cl(-) current analysis of T84 cell monolayers indicated that ISLQ blocked mainly the cystic fibrosis transmembrane conductance regulator (CFTR) Cl(-) channels, but not other unidentified cAMP-dependent Cl(-) channels. ISLQ did not affect intracellular cAMP levels or cell viability. ISLQ completely abolished the cholera toxin-induced transepithelial Cl(-) secretion in T84 cells and reduced the cholera toxin-induced intestinal fluid secretion in mouse closed loop models by 90%. Similarly, ISLQ completely inhibited the cAMP-activated apical Cl(-) current across monolayers of Madin-Darby Canine Kidney (MDCK) cells and retarded cyst growth in MDCK cyst models by 90%. This study reveals a novel action of ISLQ as a potent CFTR inhibitor with therapeutic potential for treatment of cholera and polycystic kidney disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I3766
Isoliquiritigenin, powder
C15H12O4