EMAIL THIS PAGE TO A FRIEND
International journal of dermatology
A peptide carrier for the delivery of elastin into fibroblast cells.
PMID 22788807
Abstract
Transmembrane delivery of active peptides and proteins, including skin delivery of cosmeceutical proteins such as collagen and elastin, has been a challenging issue. Amphipathic cell-penetrating peptides (CPPs) have been proposed as carrier peptides to mediate cellular uptake of proteins without covalent binding. In this study, we have used a short peptide, Pep-1, as our CPP to transport elastin into fibroblast cells. Different ratios of Pep-1/elastin complexes were produced by using a fixed amount of elastin and different molar ratio of Pep-1. The ability of transduction into cells was determined by fluorescence microscopy. The characteristics of Pep-1/elastin complexes were monitored using scanning electron microscopy and photon correlation spectroscopy. No cellular toxicity was observed in cells treated with Pep-1/elastin complex. Finally, we determined a Pep-1 : elastin ratio of 10 : 1 as the most effective ratio in cellular delivery of elastin. Pep-1 mediated fast and effective delivery of elastin as a cosmetic protein into fibroblast cells in the treatment of skin-aging symptoms.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M6500
Cysteamine hydrochloride, ≥98% (titration)
C2H7NS · HCl
30080
Cysteamine hydrochloride, ≥97.0% (RT)
C2H7NS · HCl