EMAIL THIS PAGE TO A FRIEND
Natural product communications
Anticomplement and antimicrobial activities of flavonoids from Entada phaseoloides.
PMID 22908567
Abstract
Seventeen flavonoids isolated from the extracts of the stem of Entada phaseoloides were investigated for their anticomplement (both classic and alternative pathways) and antimicrobial activities against Gram-positive bacteria (MRSA, MSSA, Standard Enterococcus and Bacillus subtilis), Gram-negative bacteria (Escherichia coli, Pseudomonas aeuroginosa) and the yeast-like pathogenic fungus Candida albicans. The anticomplement studies revealed a dose-dependent activity among isolated quercetin, luteolin, apigenin, galangin, 5,2',5'-trihydroxy-3,7,4'-trimethoxyflavone-2'-O-beta-D-glucoside (+)-3,3',5',5,7-pentahydroflavanone, (+)-dihydrokaempferol, (-)-epicatechin, (+)-catechin, naringenin, and 5,7,3',5'-tetrahydroxyflavanone, and the antimicrobial results indicated that quercetin, 5,7,4'-trihydroxy-3'-methoxyflavonol and galangin produced the inhibitory activities against MRSA, MSSA, and Standard Enterococcus, while luteolin and rhamnocitrin displayed inhibition against only MRSA and MSSA. To the best of our knowledge, this is the first report that describes the anticomplement and antimicrobial activities of the stem of E. phaseoloides.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
92204
Dihydrokaempferol, analytical standard
C15H12O6