EMAIL THIS PAGE TO A FRIEND
Journal of biological regulators and homeostatic agents
Enteropathogenic e.coli sustains iodoacetamide-induced ulcerative colitis-like colitis in rats: modulation of IL-1β, IL-6, TNF-α, COX-2, and apoptosisi.
PMID 23034271
Abstract
Pathogenic or non-pathogenic bacteria from flora may play a key role in inflammatory bowel disease (IBD) pathogenesis. However, a specific infectious agent causing IBD has not been identified. This study assessed the impact of enteropathogenic E. coli (EPEC) on the modulation of IL-1beta, IL-6, TNF- alpha, COX-2, BAX and Bcl-2 expression, in sustaining inflammation of a rat colitis model. Two hundred male Sprague-Dawley rats (4 groups) were inoculated weekly or bi-weekly for 70 days, with 1 percent methylcellulose (MC), (b) 6 percent iodoacetamide (IA) in 1 percent MC, (c) 4x108 CFU of EPEC, and (d) IA+EPEC. After a month, treatment was stopped in half of the animals in each group. IL-1beta, IL-6, TNF-alpha, COX-2, BAX and Bcl-2 expression were measured in colonic mucosa scrapings. IL-1beta, IL-6, TNF-alpha, and COX-2 were significantly increased in colonic mucosa of the IA+EPEC group and to a lesser but significant level in the IA group compared to controls, or EPEC alone, both in continued and discontinued treatment groups. Additionally, the BAX/Bcl-2 ratio decreased, indicating less apoptosis in the IA+EPEC group which exhibited more necrosis. These effects increased with experiment duration. This work provides new arguments favouring the role of bacteria in IBD pathogenesis.