EMAIL THIS PAGE TO A FRIEND
Gene
L-Carnitine intake prevents irregular feeding-induced obesity and lipid metabolism disorder.
PMID 25445284
Abstract
L-Carnitine supplementation has been used to reduce obesity caused by high-fat diet, which is beneficial for lowering blood and hepatic lipid levels, and for ameliorating fatty liver. However, whether l-carnitine may affect irregular feeding-induced obesity and lipid metabolism disorder is still largely unknown. In the present study, we developed a time-delayed pattern of eating, and investigated the effects of l-carnitine on the irregular eating induced adiposity in mice. After an experimental period of 8 weeks with l-carnitine supplementation, l-carnitine significantly inhibited body weight increase and epididymal fat weight gain induced by the time-delayed feeding. In addition, l-carnitine administration decreased levels of serum alanine aminotransferase (GPT), glutamic oxalacetic transaminase (GOT) and triglyceride (TG), which were significantly elevated by the irregular feeding. Moreover, mice supplemented with l-carnitine did not display glucose intolerance-associated hallmarks, which were found in the irregular feeding-induced obesity. Furthermore, quantitative real-time polymerase chain reaction (qRT-PCR) analysis indicated that l-carnitine counteracted the negative alterations of lipid metabolic gene expression (fatty acid synthase, 3-hydroxy-3-methyl-glutaryl coenzyme A reductase, cholesterol 7α-hydroxylase, carnitine/acylcarnitine translocase) in the liver and fat of mice caused by the irregular feeding. Therefore, our results suggest that the time-delayed pattern of eating can induce adiposity and lipid metabolic disorders, while l-carnitine supplementation might prevent these negative symptoms.