EMAIL THIS PAGE TO A FRIEND
European journal of endocrinology
Body composition, bone turnover, and bone mass in trans men during testosterone treatment: 1-year follow-up data from a prospective case-controlled study (ENIGI).
PMID 25550352
Abstract
To assess the evolution of body composition and bone metabolism in trans men during the first year of cross-sex hormonal therapy. In a prospective controlled study, we included 23 trans men (female-to-male trans persons) and 23 age-matched control women. In both groups, we examined grip strength (hand dynamometer), biochemical markers of bone turnover (C-terminal telopeptides of type 1 collagen (CTX) and procollagen 1 aminoterminal propeptide (P1NP)), total body fat and lean mass, and areal bone mineral density (aBMD) by dual-X-ray absorptiometry (DXA) and fat and muscle area at the forearm and calf, bone geometry, and volumetric bone mineral density (vBMD) by peripheral quantitative computed tomography (pQCT), before treatment and after 1 year of treatment with undecanoate (1000 mg i.m./12 weeks). Before hormonal treatment, trans men had similar bone and body composition compared with control women. Testosterone treatment induced in trans men a gain in muscle mass (+10.4%) and strength and loss of fat mass (-9.7%) (all P<0.001) and increased the levels of P1NP and CTX (both P<0.01). Areal and volumetric bone parameters remained largely unchanged apart from a small increase in trabecular vBMD at the distal radius and in BMD at the total hip in trans men (P=0.036 and P=0.001 respectively). None of these changes were observed in the control group. Short-term testosterone treatment in trans men increased muscle mass and bone turnover. The latter may rather reflect an anabolic effect of testosterone treatment rather than bone loss.