EMAIL THIS PAGE TO A FRIEND
Emerging infectious diseases
Molecular Epidemiology of Hospital Outbreak of Middle East Respiratory Syndrome, Riyadh, Saudi Arabia, 2014.
PMID 26484549
Abstract
We investigated an outbreak of Middle East respiratory syndrome (MERS) at King Fahad Medical City (KFMC), Riyadh, Saudi Arabia, during March 29-May 21, 2014. This outbreak involved 45 patients: 8 infected outside KFMC, 13 long-term patients at KFMC, 23 health care workers, and 1 who had an indeterminate source of infection. Sequences of full-length MERS coronavirus (MERS-CoV) from 10 patients and a partial sequence of MERS-CoV from another patient, when compared with other MERS-CoV sequences, demonstrated that this outbreak was part of a larger outbreak that affected multiple health care facilities in Riyadh and possibly arose from a single zoonotic transmission event that occurred in December 2013 (95% highest posterior density interval November 8, 2013-February 10, 2014). This finding suggested continued health care-associated transmission for 5 months. Molecular epidemiology documented multiple external introductions in a seemingly contiguous outbreak and helped support or refute transmission pathways suspected through epidemiologic investigation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
754935
Ethyl methyl carbonate, 99%
C4H8O3
752002
Ethyl methyl carbonate, 98%
C4H8O3