Skip to Content
MilliporeSigma

433381

Barium hexafluorosilicate

technical grade, 94%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
BaSiF6
CAS Number:
Molecular Weight:
279.40
UNSPSC Code:
12352302
PubChem Substance ID:
EC Number:
241-189-1
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

technical grade

assay

94%

form

powder

SMILES string

F[Si](F)(F)(F)(F)(F)=[Ba]



Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Codes

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library