Skip to Content
MilliporeSigma

742899

Lithium cyanide

technical, ≥96%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
CLiN
CAS Number:
Molecular Weight:
32.96
UNSPSC Code:
12352100
PubChem Substance ID:
EC Number:
219-308-3
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

technical

assay

≥96%

composition

lithium, ≥20.1%

SMILES string

[Li]C#N

InChI

1S/CN.Li/c1-2;

InChI key

KRPAPDCTWMTYHU-UHFFFAOYSA-N



Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

supp_hazards

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library