Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C9H15NO3S
CAS Number:
Molecular Weight:
217.29
EC Number:
263-607-1
UNSPSC Code:
12352200
MDL number:
Beilstein/REAXYS Number:
477887
eCl@ss:
32150313
assay
≥99.0% (HPLC)
mp
104-108 °C (lit.)
solubility
H2O: 0.1 g/mL, very slightly hazy, colorless
SMILES string
C[C@H](CS)C(=O)N1CCC[C@H]1C(O)=O
InChI
1S/C9H15NO3S/c1-6(5-14)8(11)10-4-2-3-7(10)9(12)13/h6-7,14H,2-5H2,1H3,(H,12,13)/t6-,7+/m1/s1
InChI key
FAKRSMQSSFJEIM-RQJHMYQMSA-N
Gene Information
human ... ACE(1636)
Biochem/physiol Actions
Angiotensin converting enzyme inhibitor. Inhibits the formation of angiotensin II, a bioactive peptide that stimulates angiogenesis and increases microvessel density.
Still not finding the right product?
Explore all of our products under Captopril
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Muta. 2 - Repr. 1B
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 2
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
PPE (personal protective equipment)
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
