Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C8H8
CAS Number:
Molecular Weight:
104.15
PubChem Substance ID:
UNSPSC Code:
12352103
Beilstein/REAXYS Number:
2496800
MDL number:
eCl@ss:
32160703
grade
purum
assay
≥97.0% (GC)
contains
~0.1% hydroquinone as stabilizer
refractive index
n
bp
142-143 °C (lit.)
mp
−5-−3 °C (lit.)
density
0.925 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
C1=CC=CC=CC=C1
InChI
1S/C8H8/c1-2-4-6-8-7-5-3-1/h1-8H/b2-1-,3-1-,4-2-,5-3-,6-4-,7-5-,8-6-,8-7-
InChI key
KDUIUFJBNGTBMD-BONZMOEMSA-N
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.