Skip to Content
MilliporeSigma

32389

Cefacetrile sodium

VETRANAL®, analytical standard

Synonym(s):

7-Cyanoacetylaminocephalosporanic acid sodium, Sodium (6R-trans)-3-(acetoxymethyl)-7-(cyanoacetamido)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H12N3NaO6S
CAS Number:
Molecular Weight:
361.31
UNSPSC Code:
12352200
NACRES:
NA.24
PubChem Substance ID:
EC Number:
245-513-2
Beilstein/REAXYS Number:
24235000
MDL number:
eCl@ss:
32129103
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

VETRANAL®

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

storage temp.

−20°C

SMILES string

[H][C@@]1([C@H](NC(CC#N)=O)C2=O)N2C(C([O-])=O)=C(COC(C)=O)CS1.[Na+]

InChI

1S/C13H13N3O6S.Na/c1-6(17)22-4-7-5-23-12-9(15-8(18)2-3-14)11(19)16(12)10(7)13(20)21;/h9,12H,2,4-5H2,1H3,(H,15,18)(H,20,21);/q;+1/p-1/t9-,12-;/m1./s1

InChI key

GXCRUTWHNMMJEK-WYUVZMMLSA-M

General description

Cefacetrile sodium is a synthetic cephalosporin antibiotic, which is used in the treatment of a variety of infections.

Application

Cefacetrile sodium may be used as a reference standard in the determination of cefacetrile sodium in pharmaceutical formulations using flow injection chemiluminescence (FL-CL) technique.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

WGK

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library