Skip to Content
MilliporeSigma

33898

Esprocarb

PESTANAL®, analytical standard

Synonym(s):

S-Benzyl 1,2-dimethylpropyl(ethyl)thiocarbamate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H23NOS
CAS Number:
Molecular Weight:
265.41
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
eCl@ss:
25140904
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCN(C(C)C(C)C)C(=O)SCc1ccccc1

InChI

1S/C15H23NOS/c1-5-16(13(4)12(2)3)15(17)18-11-14-9-7-6-8-10-14/h6-10,12-13H,5,11H2,1-4H3

InChI key

BXEHUCNTIZGSOJ-UHFFFAOYSA-N

General description

Esprocarb is a thiocarbamate herbicide, used for the pre- and post-emergence control of annual weeds and branyard grass in rice.

Application

Esprocarb may be used as an analytical reference standard for the determination of the analyte in food samples using solid phase extraction combined with gas chromatography coupled to mass spectrometry (SPE-GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


Pictograms

Environment

Hazard Codes

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class

10 - Combustible liquids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library