Skip to Content
MilliporeSigma

34525

Flusulfamide

PESTANAL®, analytical standard

Synonym(s):

2′,4-Dichloro-α,α,α-trifluoro-4′-nitro-m-toluenesulfonanilide, 4-Chloro-N-(2-chloro-4-nitrophenyl)-3-(trifluoromethyl)benzenesulfonamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H7Cl2F3N2O4S
CAS Number:
Molecular Weight:
415.17
PubChem Substance ID:
UNSPSC Code:
12352202
Beilstein/REAXYS Number:
11322439
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

O=S(NC1=C(Cl)C=C([N+]([O-])=O)C=C1)(C2=CC(C(F)(F)F)=C(Cl)C=C2)=O

InChI

1S/C13H7Cl2F3N2O4S/c14-10-3-2-8(6-9(10)13(16,17)18)25(23,24)19-12-4-1-7(20(21)22)5-11(12)15/h1-6,19H

InChI key

GNVDAZSPJWCIQZ-UHFFFAOYSA-N

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Codes

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library