Skip to Content
MilliporeSigma

56528

Bisphenol PH

analytical standard

Synonym(s):

2,2-Bis(2-hydroxy-5-biphenylyl)propane, 2,2-Bis(4-hydroxy-3-phenylphenyl)propane, 2,2-{5,5′-Bis[1,1′-(biphenyl)-2-ol]}propane, 5,5′-(1-Methylethylidene)bis[(1,1′-biphenyl)-2-ol], 5,5′-Isopropylidenedi-2-biphenylol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C27H24O2
CAS Number:
Molecular Weight:
380.48
PubChem Substance ID:
UNSPSC Code:
12352200
Beilstein/REAXYS Number:
5117041
MDL number:
eCl@ss:
32129103
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

format

neat

SMILES string

OC1=CC=C(C(C)(C)C2=CC(C3=CC=CC=C3)=C(O)C=C2)C=C1C4=CC=CC=C4

InChI

1S/C27H24O2/c1-27(2,21-13-15-25(28)23(17-21)19-9-5-3-6-10-19)22-14-16-26(29)24(18-22)20-11-7-4-8-12-20/h3-18,28-29H,1-2H3

InChI key

BKTRENAPTCBBFA-UHFFFAOYSA-N



Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

13 - Non Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library