Select a Size
Change View
About This Item
Linear Formula:
C6H5MgBr
CAS Number:
Molecular Weight:
181.31
UNSPSC Code:
12352103
Beilstein/REAXYS Number:
3588849
MDL number:
eCl@ss:
32150313
grade
purum
concentration
~3 M in diethyl ether
density
1.13 g/mL at 20 °C
SMILES string
Br[Mg]c1ccccc1
InChI
1S/C6H5.BrH.Mg/c1-2-4-6-5-3-1;;/h1-5H;1H;/q;;+1/p-1
InChI key
ANRQGKOBLBYXFM-UHFFFAOYSA-M
Still not finding the right product?
Explore all of our products under Phenylmagnesium bromide solution
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3
Target Organs
Central nervous system
supp_hazards
Storage Class
3 - Flammable liquids
WGK
WGK 3
Flash Point (°F)
-40.0 °F - closed cup
Flash Point (°C)
-40 °C - closed cup
PPE (personal protective equipment)
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


