Skip to Content
MilliporeSigma

79225

4-Phenyl-1,2,4-triazoline-3,5-dione

purum, ≥97.0% (N)

Synonym(s):

4-Phenyl-3H-1,2,4-triazole-3,5(4H)-dione, PTAD

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C8H5N3O2
CAS Number:
Molecular Weight:
175.14
EC Number:
224-191-7
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
141548
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum

assay

≥97.0% (N)

mp

165-170 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

O=C1N=NC(=O)N1c2ccccc2

InChI

1S/C8H5N3O2/c12-7-9-10-8(13)11(7)6-4-2-1-3-5-6/h1-5H

InChI key

ISULLEUFOQSBGY-UHFFFAOYSA-N

Application

The most reactive dienophile known


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library