Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C18H16O7
CAS Number:
Molecular Weight:
344.32
PubChem Substance ID:
UNSPSC Code:
12352000
EC Number:
231-456-0
MDL number:
Beilstein/REAXYS Number:
96698
assay
≥95.0% (sum of enantiomers, TLC)
optical activity
[α]20/D +495±15°, c = 0.7% in chloroform
mp
194-204 °C
antibiotic activity spectrum
fungi
mode of action
enzyme | inhibits
SMILES string
CC(=O)c1c(O)c(C)c(O)c2c1OC3=CC(=O)C(C(C)=O)=C(O)[C@@]23C
General description
Chemical structure: polyphenolic acid
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.