Skip to Content
MilliporeSigma

Skip To

95740

DL-Xylose

purum, ≥99.0% (HPLC)

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
UNSPSC Code:
12352201
EC Number:
247-395-8
Beilstein/REAXYS Number:
1723087
MDL number:


grade

purum

assay

≥99.0% (HPLC)

mp

133-136 °C

solubility

H2O: 0.1 g/mL, clear, colorless

SMILES string

OC[C@@H](O)[C@H](O)[C@@H](O)C=O



Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library