Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
lyophilized powder
mol wt
18 kDa
species reactivity
guinea pig, human, bovine, mouse, horse, canine, rat
concentration
0.5 mg - 1 mg/mL
technique(s)
western blot: suitable
NCBI accession no.
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... GADD45B(4616)
General description
Growth arrest and DNA-damage-inducible, β (GADD45B) regulates DNA demethylation, adult neurogenesis, p38 signaling and cellular apoptosis. Genetic defects or lack of GADD45B are known to affect long-term memory and synaptic plasticity.
Rabbit Anti-GADD45B antibody recognizes bovine, human, mouse, rat, and canine GADD45B.
Rabbit Anti-GADD45B antibody recognizes bovine, human, mouse, rat, and canine GADD45B.
Immunogen
The immunogen for anti-GADD45B antibody: synthetic peptide derected towards the middle region of human GADD45B
Application
Rabbit Anti-GADD45B antibody is suitable for western blot applications at a concentration of 0.25 μg/ml.
Physical form
Lyophilized from PBS buffer with 2% sucrose
Other Notes
Synthetic peptide located within the following region: FCCDNDINIVRVSGMQRLAQLLGEPAETQGTTEARDLHCLLVTNPHTDAW
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.