Skip to Content
MilliporeSigma

F3137

Fast Orange GR Salt

Synonym(s):

2-Nitrobenzenediazonium chloride hemi(zinc chloride) salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H4N3O2 · 1/2ZnCl4
CAS Number:
Molecular Weight:
253.72
Colour Index Number:
37025
EC Number:
239-712-3
UNSPSC Code:
12171500
eCl@ss:
32150313
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


composition

Dye content, ~90%

storage temp.

−20°C





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library