Skip to Content
MilliporeSigma

SAB2500331

Anti-DUSP1 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CL100, Anti-Dual specificity phosphatase 1, Anti-HVH1, Anti-MKP-1, Anti-PTPN10

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ELISA (i), IHC, WB
Citations:
11
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

technique(s)

immunohistochemistry: suitable, indirect ELISA: suitable, western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DUSP1(1843)

General description

Dual specificity phosphatase 1 (DUSP1) is a protein phosphatase. The gene encoding it is localized on human chromosome 5q35.1.

Immunogen

Peptide with sequence SYLQSPITTSPSC from the C Terminus of the protein sequence according to NP_004408.1.

Application

Anti-DUSP1 antibody produced in goat has been used for Western blotting.

Biochem/physiol Actions

Dual specificity phosphatase 1 (DUSP1) modulates the activity of mitogen-activated protein kinase (MAPK). It targets and dephosphorylates extracellular signal-regulated kinases 1 and 2 (ERK 1/2). During cellular responses to oxidative stress, the activity of DUSP1 is regulated by p53. Overexpression of the protein has been associated with various tumors.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library