Skip to Content
MilliporeSigma

E-003

Ecgonine-d3 hydrochloride

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Ecgonine-D3 hydrochloride solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H12NO3D3·HCl
CAS Number:
Molecular Weight:
224.70
UNSPSC Code:
41116107
EC Number:
200-659-6
eCl@ss:
25140904
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IB (Portugal)

concentration

100 μg/mL in methanol (as free base)

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

2-8°C

InChI

1S/C9H15NO3.ClH/c1-10-5-2-3-6(10)8(9(12)13)7(11)4-5;/h5-8,11H,2-4H2,1H3,(H,12,13);1H/i1D3;

InChI key

HVWQTEPEBQYIFB-NIIDSAIPSA-N

General description

Ecgonine-D3 HCl is a stable-labeled internal standard suitable for GC/MS or LC/MS testing applications in clinical toxicology, isotope dilution methods, forensic analysis or urine drug testing. Ecgonine is not only a precursor of cocaine, but also a metabolite obtained by cocaine hydrolysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class

3 - Flammable liquids

WGK

WGK 1

Flash Point (°F)

49.5 °F - closed cup

Flash Point (°C)

9.7 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library