Custom Peptides
- High quality peptide synthesis using FMOC and/or BOC chemistries
- Sequence length: 2-40mer
- Amount: up to 1 gram
- Purity Levels: Unpurified, 70%, 80%, 95%
- Comprehensive Modification offering including, FRET, C-terminal aldehyde, phosphorylation, multiple disulfide bonds, etc.
- Format: Supplied lyophilized
- QC Analysis: All peptides by HPLC and MS; additional analysis available upon request
- Learn more
Sign In To Continue
Don't Have An Account?Articles
Consider sequence length, solubility, and stability when designing peptides for custom synthesis.
Related Content
Peptide synthesis up to 200 amino acids with HPLC purification, offering various purity levels, scales, and modifications.
Magdalena Banas et al.
PloS one, 8(3), e58709-e58709 (2013-03-26)
Chemerin, a chemoattractant ligand for chemokine-like receptor 1 (CMKLR1) is predicted to share similar tertiary structure with antibacterial cathelicidins. Recombinant chemerin has antimicrobial activity. Here we show that endogenous chemerin is abundant in human epidermis, and that inhibition of bacteria
Artur Schmidtchen et al.
The Journal of biological chemistry, 284(26), 17584-17594 (2009-04-29)
A novel approach for boosting antimicrobial peptides through end tagging with hydrophobic oligopeptide stretches is demonstrated. Focusing on two peptides derived from kininogen, GKHKNKGKKNGKHNGWK (GKH17) and HKHGHGHGKHKNKGKKN (HKH17), tagging resulted in enhanced killing of Gram-positive Staphylococcus aureus, Gram-negative Escherichia coli
Joyce J C Kroot et al.
American journal of hematology, 87(10), 977-983 (2012-08-14)
Measurements of the iron regulatory hormone hepcidin by various methodologies and laboratories are not harmonized. As a result different numeric results are obtained for the same clinical sample. We investigated whether better agreement between plasma hepcidin methods can be achieved
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 208902-500UG | 04055977219500 |
| 883061 | 04053252942792 |
| 916110-1EA | 04061842192915 |
| 12-355 | 04053252474682 |
| 11666975001 | 04061838704542 |
| 883060 | 04053252942785 |
| V7754-4MG | 04061837481369 |
| P4480-1MG | 04061823013833 |
| 12-123 | 04053252509568 |
| 12-347 | 04053252334504 |
| 208735-500UG | 04055977202779 |
| SRP6562-20UG | 04061837311543 |
| P5801-.5MG | 04061832677668 |
| P0240-200UG | 04061838071781 |
| SRP0400-80NMOL | 04061837197062 |
| AG992 | 04053252279133 |
| 12-357 | 04053252362101 |
| 12-459 | 04053252736889 |
| 914185-1EA | 04061842072385 |