Skip to Content
MilliporeSigma

697524

Strontium tetramethylheptanedionate

anhydrous, 99.9% trace metals basis

Synonym(s):

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)strontium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H38O4Sr
CAS Number:
Molecular Weight:
454.15
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

anhydrous

product line

Sol-Gel Precursors

assay

99.9% trace metals basis

form

solid

reaction suitability

core: strontium

mp

210 °C

SMILES string

[Sr++].CC(C)(C)C(\[O-])=C\C(=O)C(C)(C)C.CC(C)(C)C(\[O-])=C\C(=O)C(C)(C)C

InChI

1S/2C11H20O2.Sr/c2*1-10(2,3)8(12)7-9(13)11(4,5)6;/h2*7,12H,1-6H3;/q;;+2/p-2/b2*8-7-;

InChI key

WQTMVGGQTAPQJQ-ATMONBRVSA-L

General description

Atomic number of base material: 38 Strontium


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library