Skip to Content
MilliporeSigma

725536

Tetrakis(diethylamido)titanium(IV)

packaged for use in deposition systems

Synonym(s):

TDEAT, Tetrakis(diethylamino)titanium(IV), Titanium(IV) tetrakis(diethylamide)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
[(C2H5)2N]4Ti
CAS Number:
Molecular Weight:
336.38
UNSPSC Code:
12352103
NACRES:
NA.23
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
3621431
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

reaction suitability

core: titanium, reagent type: catalyst

refractive index

n20/D 1.536 (lit.)

bp

112 °C/0.1 mmHg (lit.)

density

0.931 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)[Ti](N(CC)CC)(N(CC)CC)N(CC)CC

InChI

1S/4C4H10N.Ti/c4*1-3-5-4-2;/h4*3-4H2,1-2H3;/q4*-1;+4

InChI key

VJDVOZLYDLHLSM-UHFFFAOYSA-N

General description

Atomic number of base material: 22 Titanium


Still not finding the right product?

Explore all of our products under Tetrakis(diethylamido)titanium(IV)


Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Codes

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point (°F)

-19.8 °F - closed cup

Flash Point (°C)

-28.8 °C - closed cup

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B - Water-react. 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library