Skip to Content
MilliporeSigma

744670

Bis(5H-dibenzo[a,d]cyclohepten-5-yl)amine

97%

Synonym(s):

trop2-NH

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C30H23N
CAS Number:
Molecular Weight:
397.51
UNSPSC Code:
12352300
NACRES:
NA.22
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
2778898
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥96.5% (HPLC), 97%

form

powder

reaction suitability

reagent type: ligand

SMILES string

C1(C(C=CC=C2)=C2C=CC3=C1C=CC=C3)NC4C(C=CC=C5)=C5C=CC6=C4C=CC=C6

InChI

1S/C30H23N/c1-5-13-25-21(9-1)17-18-22-10-2-6-14-26(22)29(25)31-30-27-15-7-3-11-23(27)19-20-24-12-4-8-16-28(24)30/h1-20,29-31H

InChI key

OFFKQRRCBWRRFP-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Hazard Codes

Hazard Classifications

Aquatic Chronic 4

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library