Skip to Content
MilliporeSigma

746533

Poly(L-lactide) 4-cyano-4-[(dodecylsulfanylthiocarbonyl)sulfanyl]pentonate

Mn 10,000, PDI ≤1.2

Synonym(s):

PLA RAFT, PLLA RAFT, PLLA thiocarbonyl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C19H35NOS3(C3H4O2)n
NACRES:
NA.23
UNSPSC Code:
12162002
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

mol wt

Mn 10,000

mp

>300 °C

PDI

≤1.2

storage temp.

2-8°C

Application

This Poly(L-lactide) is a biodegradable polymer that contains RAFT agent functionality. The benzodithioate moiety is well suited to polymerize methacrylates and methacrylamides. PLA is commonly used in several applications including drug delivery, nanoparticle surface modification, end-group functionalization, macroinitiator or block precursor.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library