Skip to Content
MilliporeSigma

769398

Hafnium(IV) oxide

pellets, diam. × thickness 9 mm × 4.2 mm

Synonym(s):

Hafnia

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
HfO2
CAS Number:
Molecular Weight:
210.49
UNSPSC Code:
12352200
NACRES:
NA.23
EC Number:
235-013-2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

pellets

diam. × thickness

9 mm × 4.2 mm

impurities

≤0.1% PBB (polybrominated biphenyls), ≤0.1% PBDE (polybrominated diphenyl ethers)

density

9.68 g/mL at 25 °C (lit.)

cation traces

Cd: ≤0.01%, Co: ≤0.0005%, Cr: ≤0.005%, Cu: ≤0.0005%, Fe: ≤0.005%, Hg: ≤0.1%, Pb: ≤0.1%, Ti: ≤0.005%, V: ≤0.005%, Zr: ≤1.0%

SMILES string

O=[Hf]=O

InChI

1S/Hf.2O

InChI key

CJNBYAVZURUTKZ-UHFFFAOYSA-N

Application

High dielectric constant of hafnia makes it suitable for application in metal oxide based semiconductor devices. It also finds applications as optical coating materials.


Still not finding the right product?

Explore all of our products under Hafnium(IV) oxide


Storage Class

11 - Combustible Solids

WGK

nwg

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library