Skip to Content
MilliporeSigma

790265

Poly(D,L-lactide-co-glycolide)-block-poly(ethylene glycol)-block-poly(D,L-lactide-co-glycolide) based poly(ether ester urethane)

average Mn (8,000-20,000)

Synonym(s):

PEEU, PLGA-PEG-PLGA urethane, VISCOPRENE® 2, poly(ester ether urethane)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
[C2H2N2O2R[(C3H4O2)x(C2H2O2)y]m[C2H4O]lO[(C3H4O2)x(C2H2O2)y]m]n
UNSPSC Code:
12352100
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid (or solid)

feed ratio

19:(65:16) (PEG:(DL-Lactide:Glycolide)), lactide:glycolide 80:20

mol wt

PEG average Mn 400, average Mn (8,000-20,000)

transition temp

Tg <0 °C

PDI

<2.25

storage temp.

2-8°C

SMILES string

CC(N[R]NC(OC(C)C(OCC(OCCOC(C(C)OC(COC)=O)=O)=O)=O)=O)=O.CC(N[R]NC(OCCCOC)=O)=O

Legal Information

Product of Poly-Med, Inc. VISCOPRENE® is a registered trademark of Poly-Med, Inc.
Viscoprene is a registered trademark of Poly-Med, Inc.



Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library