Skip to Content
MilliporeSigma

791067

Polystyrene, amine terminated

average Mn 10,000, PDI ≤1.3

Synonym(s):

PS NH2, amino PS, aminopolystyrene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(C8H8)nCH5N
NACRES:
NA.23
UNSPSC Code:
12352100
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

mol wt

average Mn 10,000

feature

hydrophobic

PDI

≤1.3

Application

Functional polystyrene with amine termination has been used as a macromolecular surfactant; capable of conjugation with enzymes; proteins and lipases. They are also known to form well-defined assemblies in aqueous solution.

Legal Information

Sold for research purposes only and subject to field restriction: see www.sigmaaldrich.com/raftlicense. Patents: 7; 714; 075; 7; 250; 479; 7; 666; 962; 6; 642; 318; 6; 747; 111; W2010/8356.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library