Skip to Content
MilliporeSigma

806528

Ac-Thr(t-Bu)-OH

Synonym(s):

(S)-3-tert-Butoxy-2-(acylamino)butyric acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H19NO4
CAS Number:
Molecular Weight:
217.26
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

95%

form

powder

mp

137 °C

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

O=C(O)[C@H]([C@@H](OC(C)(C)C)C)NC(C)=O

InChI

1S/C10H19NO4/c1-6(15-10(3,4)5)8(9(13)14)11-7(2)12/h6,8H,1-5H3,(H,11,12)(H,13,14)/t6?,8-/m0/s1

InChI key

JTQQJUFULLOMEJ-XDKWHASVSA-N

General description

Ac-Thr(t-Bu)-OH is a threonine derivative.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library