Skip to Content
MilliporeSigma

900318

Poly(L-lactide-co-trimethylene carbonate)

lactide:TMC 20:80, viscosity 1.5 dL/g 

Synonym(s):

PLA-TMC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3[C6H8O4]m[C4H6O3]nCH3
NACRES:
NA.23
UNSPSC Code:
12162002
eCl@ss:
32160703
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

chips or granules (solid)

feed ratio

lactide:trimethylene carbonate 20:80

viscosity

1.5 dL/g

storage temp.

−20°C



Still not finding the right product?


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library