Skip to Content
MilliporeSigma

900488

Gold nano-urchins

90 nm, maleimide functionalized, conjugation kit

Synonym(s):

Maleimide-activated gold nanourchins, Au nanoparticles, Gold nanoparticles, Urchins

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.23
UNSPSC Code:
12141717
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

nanourchin

shelf life

12 mo. at -20 °C (If stored unopened and as specified, are stable for at least 12 months)

storage condition

protect from light

particle size

90 nm

λmax

630 nm

functional group

maleimide

Legal Information

Product of CytoDiagnostics, Inc.


Still not finding the right product?

Explore all of our products under Gold nano-urchins


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library