Skip to Content
MilliporeSigma

900491

Gold nano-urchins

60 nm, NHS ester functionalized, conjugation kit

Synonym(s):

NHS-activated gold nanourchins, Au nanoparticles, Gold nanoparticles, Urchins

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.23
UNSPSC Code:
12141717
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

nanourchin

shelf life

12 mo. at -20 °C (If stored unopened and as specified, are stable for at least 12 months)

storage condition

protect from light

particle size

60 nm

λmax

585 nm

functional group

NHS ester

storage temp.

−20°C

Legal Information

Product of CytoDiagnostics, Inc.


Still not finding the right product?

Explore all of our products under Gold nano-urchins


Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library